missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dihydro-7-azaindole 98.0+%, TCI America™

Catalog No. D50781G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D50781G 1 g
D50785G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D50781G Supplier TCI America Supplier No. D50781G

Chemical Identifiers

CAS 10592-27-5
Molecular Formula C7H8N2
Molecular Weight (g/mol) 120.155
MDL Number MFCD06659751
InChI Key ZFFYPGZDXUPKNK-UHFFFAOYSA-N
Synonym 2,3-dihydro-1h-pyrrolo 2,3-b pyridine, 7-azaindoline, 2,3-dihydro-7-azaindole, 1h,2h,3h-pyrrolo 2,3-b pyridine, 1h-pyrrolo 2,3-b pyridine, 2,3-dihydro, 2,3-dihydro-1h-pyrrolo 2,3-b pyridine 7-azaindoline, pubchem18022, acmc-2098hy, ksc492q5f
PubChem CID 10011889
IUPAC Name 2,3-dihydro-1H-pyrrolo[2,3-b]pyridine
SMILES C1CNC2=C1C=CC=N2

Specifications

Melting Point 83°C
Boiling Point 114°C
Quantity 1 g
Formula Weight 120.16
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Dihydro-7-azaindole