missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dihydro-7-azaindole, 97+%

Catalog No. AAH6455114
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6455114 25 g
AAH6455106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6455114 Supplier Thermo Scientific Chemicals Supplier No. H6455114
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 10592-27-5 | C7H8N2 | 120.155 g/mol

It can be used as pharmaceutical raw material and intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It can be used as pharmaceutical raw material and intermediate.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.

Chemical Identifiers

CAS 10592-27-5
Molecular Formula C7H8N2
Molecular Weight (g/mol) 120.155
MDL Number MFCD06659751
InChI Key ZFFYPGZDXUPKNK-UHFFFAOYSA-N
Synonym 2,3-dihydro-1h-pyrrolo 2,3-b pyridine, 7-azaindoline, 2,3-dihydro-7-azaindole, 1h,2h,3h-pyrrolo 2,3-b pyridine, 1h-pyrrolo 2,3-b pyridine, 2,3-dihydro, 2,3-dihydro-1h-pyrrolo 2,3-b pyridine 7-azaindoline, pubchem18022, acmc-2098hy, ksc492q5f
PubChem CID 10011889
IUPAC Name 2,3-dihydro-1H-pyrrolo[2,3-b]pyridine
SMILES C1CNC2=C1C=CC=N2

Specifications

Melting Point 78°C to 83°C
Color White
Quantity 25 g
Beilstein 116366
Solubility Information Slightly soluble in water.
Formula Weight 120.15
Percent Purity ≥97%
Physical Form Powder
Chemical Name or Material 2,3-Dihydro-7-azaindole
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only