missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dihydro-2-spiro-7'-[8'-imino-7',8'-dihydronaphthalen-1'-amine]perimidine, TCI America™

Catalog No. D3619100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D3619100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D3619100MG Supplier TCI America Supplier No. D3619100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 851768-62-2
Molecular Formula C20H16N4
Molecular Weight (g/mol) 312.376
InChI Key CXGACCZEFCWAJN-UHFFFAOYSA-N
Synonym 1-Iminospiro[naphthalene-2(1H),2′C(3′CH)-[1H]perimidin]-8-amine, PNI
PubChem CID 44629896
IUPAC Name 8'-iminospiro[1,3-dihydroperimidine-2,7'-naphthalene]-1'-amine
SMILES C1=CC2=C(C(=C1)N)C(=N)C3(C=C2)NC4=CC=CC5=C4C(=CC=C5)N3

Specifications

Quantity 100 mg
Formula Weight 312.38
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Dihydro-2-spiro-7′-[8′-imino-7′,8′-dihydronaphthalen-1′-amine]peri
TSCA No
Recommended Storage Freezer