missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dihydro-1-benzofuran-5-ylboronic acid, 95%, May contain varying amounts of anhydride, Thermo Scientific™

Catalog No. p-2696927
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC00812DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC00812DA Supplier Thermo Scientific Chemicals Supplier No. CC00812DA

Chemical Identifiers

CAS 227305-69-3
Molecular Formula C8H9BO3
Molecular Weight (g/mol) 163.97
MDL Number MFCD02681979
InChI Key ZIXLJHSFAMVHPC-UHFFFAOYSA-N
Synonym 2,3-dihydrobenzofuran-5-boronic acid, 2,3-dihydrobenzofuran-5-ylboronic acid, 2,3-dihydrobenzofuran-5-yl boronic acid, 2,3-dihydrobenzo b furan-5-boronic acid, 2,3-dihydrobenzofuran-5-yl-5-boronic acid, boronic acid, 2,3-dihydro-5-benzofuranyl, 2,3-dihydro-1-benzofuran-5-yl-boranediol, 2,3-dihydro-1-benzofuran-5-yl boronic acid, pubchem7848, zlchem 1108
PubChem CID 2773380
SMILES OB(O)C1=CC=C2OCCC2=C1

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber glass bottle
Quantity 1 g
IUPAC Name 2,3-dihydro-1-benzofuran-5-ylboronic acid
Formula Weight 163.97
Percent Purity 95%
Chemical Name or Material 2, 3-Dihydro-1-benzofuran-5-ylboronic acid
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.