missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dihydro-1,4-benzodioxine-6-carboxylic acid, 97%, Thermo Scientific™

Catalog No. CC01301CB
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC01301CB 250 mg
CC01301DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC01301CB Supplier Thermo Scientific Chemicals Supplier No. CC01301CB

Chemical Identifiers

CAS 4442-54-0
Molecular Formula C9H8O4
Molecular Weight (g/mol) 180.159
MDL Number MFCD00463509
InChI Key JWZQJTGQFHIRFQ-UHFFFAOYSA-N
Synonym 1,4-benzodioxane-6-carboxylic acid, 2,3-dihydrobenzo b 1,4 dioxine-6-carboxylic acid, 3,4-ethylenedioxy benzoic acid, 2,3-dihydro-benzo 1,4 dioxine-6-carboxylic acid, 1,4-benzodioxin-6-carboxylic acid, 2,3-dihydro, 3,4-ethylenedioxybenzoic acid, 1,4-benzodioxan-6-carboxylic acid, 2,3-dihydro-1,4-benzodioxin-6-carboxylic acid, 2,3-dihydro-1,4-benzodioxine-6-carboxylicacid, 6-carboxy-2,3-dihydro-1,4-benzodioxine
PubChem CID 2758833
IUPAC Name 2,3-dihydro-1,4-benzodioxine-6-carboxylic acid
SMILES C1COC2=C(O1)C=CC(=C2)C(=O)O

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 180.16
Percent Purity 95%
Chemical Name or Material 2, 3-Dihydro-1, 4-benzodioxine-6-carboxylic acid