missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dihydro-1,4-benzodioxine-6-carbaldehyde 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC01304DE

Quantity 5g

 View more versions of this product

Catalog No. CC01304DE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

29668-44-8
164.16
1,4-benzodioxan-6-carboxaldehyde, 1,4-benzodioxan-6-carbaldehyde, 2,3-dihydrobenzo b 1,4 dioxine-6-carbaldehyde, 1,4-benzodioxane-6-carboxaldehyde, 3,4-ethylenedioxybenzaldehyde, 1,4-benzodioxin-6-carboxaldehyde, 2,3-dihydro, 7-formyl-1,4-benzodioxane, 1,4-benzodioxane-6-aldehyde, 2,3-dihydro-1,4-benzodioxin-6-carbaldehyde, 1,4-benzodioxane-6-carbaldehyde
2,3-dihydro-1,4-benzodioxine-6-carbaldehyde
C9H8O3
CWKXDPPQCVWXAG-UHFFFAOYSA-N
248127
C1COC2=C(O1)C=CC(=C2)C=O

Specifications

2, 3-Dihydro-1, 4-benzodioxine-6-carbaldehyde
97%
5g
CWKXDPPQCVWXAG-UHFFFAOYSA-N
2,3-dihydro-1,4-benzodioxine-6-carbaldehyde
248127
97%
29668-44-8
C9H8O3
1,4-benzodioxan-6-carboxaldehyde, 1,4-benzodioxan-6-carbaldehyde, 2,3-dihydrobenzo b 1,4 dioxine-6-carbaldehyde, 1,4-benzodioxane-6-carboxaldehyde, 3,4-ethylenedioxybenzaldehyde, 1,4-benzodioxin-6-carboxaldehyde, 2,3-dihydro, 7-formyl-1,4-benzodioxane, 1,4-benzodioxane-6-aldehyde, 2,3-dihydro-1,4-benzodioxin-6-carbaldehyde, 1,4-benzodioxane-6-carbaldehyde
C1COC2=C(O1)C=CC(=C2)C=O
164.16
164.16
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents