missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dihydro-1,4-benzodioxine-5-carboxamide, 97%, Thermo Scientific™

Catalog No. p-2696882
Change view
Click to view available options
Quantity:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MO00789DA 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MO00789DA Supplier Thermo Scientific Chemicals Supplier No. MO00789DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 349550-81-8
Molecular Formula C9H9NO3
Molecular Weight (g/mol) 179.175
MDL Number MFCD03659722
InChI Key OIYLTYTXMKZKCR-UHFFFAOYSA-N
Synonym 2,3-dihydrobenzo b 1,4 dioxine-5-carboxamide, maybridge3_004369, 2h,3h-benzo e 1,4-dioxane-5-carboxamide, 1,4-benzodioxin-5-carboxamide,2,3-dihydro, 5wy
PubChem CID 2779845
IUPAC Name 2,3-dihydro-1,4-benzodioxine-5-carboxamide
SMILES C1COC2=C(C=CC=C2O1)C(=O)N

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1g
Formula Weight 179.17
Percent Purity 97%
Chemical Name or Material 2, 3-Dihydro-1, 4-benzodioxine-5-carboxamide