missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Dihydro-1,4-benzodioxine-5-carbaldehyde, Thermo Scientific™
Specifications
Specifications
| Assay Percent Range | 97% |
| Packaging | Amber Glass Bottle |
| Quantity | 5g |
| Formula Weight | 164.16 |
| Percent Purity | 97% |
| Chemical Name or Material | 2, 3-Dihydro-1, 4-benzodioxine-5-carbaldehyde |