missing translation for 'onlineSavingsMsg'
Learn More

2',3'-Difluoroacetophenone, 97%

Catalog No. p-7023344
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1678606 5 g
AAA1678614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1678606 Supplier Thermo Scientific Chemicals Supplier No. A1678606

CAS: 18355-80-1 | C8H6F2O | 156.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 18355-80-1
Molecular Formula C8H6F2O
Molecular Weight (g/mol) 156.13
MDL Number MFCD00061274
InChI Key PQUXFUBNSYCQAL-UHFFFAOYSA-N
Synonym 2',3'-difluoroacetophenone, 1-2,3-difluorophenyl ethanone, 2,3-difluoroacetophenone, 1-2,3-difluorophenyl ethan-1-one, ethanone, 1-2,3-difluorophenyl, 1-acetyl-2,3-difluorobenzene, pubchem4211, acmc-209x0g, ksc184c5n, 2',3'-difluoro acetophenone
PubChem CID 519550
SMILES CC(=O)C1=CC=CC(F)=C1F

Specifications

Boiling Point 84°C to 85°C (20 mmHg)
Odor Characteristic
Refractive Index 1.489
Quantity 5 g
Beilstein 2963572
IUPAC Name 1-(2,3-difluorophenyl)ethanone
Formula Weight 156.13
Percent Purity 97%
Chemical Name or Material 2',3'-Difluoroacetophenone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only