missing translation for 'onlineSavingsMsg'
Learn More

2,3-Difluoro-6-nitrophenylacetic Acid 98.0+%, TCI America™

Catalog No. p-7128352
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D45831G 1 g
D45835G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D45831G Supplier TCI America Supplier No. D45831G
Only null left

Chemical Identifiers

CAS 141428-47-9
Molecular Formula C8H5F2NO4
Molecular Weight (g/mol) 217.128
MDL Number MFCD11053839
InChI Key KZNVCGPCWKYPKD-UHFFFAOYSA-N
PubChem CID 22475350
IUPAC Name 2-(2,3-difluoro-6-nitrophenyl)acetic acid
SMILES C1=CC(=C(C(=C1[N+](=O)[O-])CC(=O)O)F)F

Specifications

Melting Point 156°C
Color Yellow
Quantity 1 g
Formula Weight 217.13
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Difluoro-6-nitrophenylacetic Acid