missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Difluoro-4-methoxyphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
Supplier: TCI America D50165G
Specifications
| 2,3-Difluoro-4-methoxyphenylboronic Acid (contains varying amounts of Anhydride) | |
| White-Yellow | |
| MFCD07368234 | |
| 2,3-difluoro-4-methoxyphenyl boronic acid, 2,3-difluoro-4-methoxybenzeneboronic acid, 2,3-difluoro-4-methyloxyphenylboronic acid, boronic acid, 2,3-difluoro-4-methoxyphenyl, boronic acid, b-2,3-difluoro-4-methoxyphenyl, 2,3-difluoro-4-methoxyphenylboronicacid, 4-borono-2,3-difluoroanisole, 2,3-difluoroanisylboronic acid, 2,3-difluoro-4-methoxyphenylboronic aicd, 2,3-difluoro-4-methoxyphenyl boronicacid | |
| COC1=C(F)C(F)=C(C=C1)B(O)O | |
| 187.94 | |
| 187.94 |
| 170981-41-6 | |
| C7H7BF2O3 | |
| 5 g | |
| HIWYFMBSFSTYGU-UHFFFAOYSA-N | |
| (2,3-difluoro-4-methoxyphenyl)boronic acid | |
| 25067272 | |
| Crystalline Powder |
Chemical Identifiers
| 170981-41-6 | |
| MFCD07368234 | |
| 25067272 |
| C7H7BF2O3 | |
| HIWYFMBSFSTYGU-UHFFFAOYSA-N | |
| (2,3-difluoro-4-methoxyphenyl)boronic acid |
Safety and Handling
TSCA : No