missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Diethyl-5-methylpyrazine 97.0+%, TCI America™
$113.71 - $303.84
Chemical Identifiers
| CAS | 18138-04-0 |
|---|---|
| Molecular Formula | C9H14N2 |
| Molecular Weight (g/mol) | 150.23 |
| MDL Number | MFCD00009635 |
| InChI Key | PSINWXIDJYEXLO-UHFFFAOYSA-N |
| Synonym | pyrazine, 2,3-diethyl-5-methyl, 2-methyl-5,6-diethylpyrazine, 2,3-diethyl-6-methylpyrazine, 5-methyl-2,3-diethylpyrazine, 2,3-diethyl-5-methyl-pyrazine, fema no. 3336, hazelnut pyrazine, pubchem8621, acmc-1ca9d, ksc491g2h |
| PubChem CID | 28905 |
| IUPAC Name | 2,3-diethyl-5-methylpyrazine |
| SMILES | CCC1=NC=C(C)N=C1CC |
Chemical Identifiers
| 18138-04-0 | |
| 150.23 | |
| PSINWXIDJYEXLO-UHFFFAOYSA-N | |
| 28905 | |
| CCC1=NC=C(C)N=C1CC |
| C9H14N2 | |
| MFCD00009635 | |
| pyrazine, 2,3-diethyl-5-methyl, 2-methyl-5,6-diethylpyrazine, 2,3-diethyl-6-methylpyrazine, 5-methyl-2,3-diethylpyrazine, 2,3-diethyl-5-methyl-pyrazine, fema no. 3336, hazelnut pyrazine, pubchem8621, acmc-1ca9d, ksc491g2h | |
| 2,3-diethyl-5-methylpyrazine |
Specifications
| 18138-04-0 | |
| 80°C | |
| MFCD00009635 | |
| 1993 | |
| PSINWXIDJYEXLO-UHFFFAOYSA-N | |
| 2,3-diethyl-5-methylpyrazine | |
| 28905 | |
| ≥97.0% (GC) | |
| 2,3-Diethyl-5-methylpyrazine |
| Yellow | |
| C9H14N2 | |
| 5 mL | |
| pyrazine, 2,3-diethyl-5-methyl, 2-methyl-5,6-diethylpyrazine, 2,3-diethyl-6-methylpyrazine, 5-methyl-2,3-diethylpyrazine, 2,3-diethyl-5-methyl-pyrazine, fema no. 3336, hazelnut pyrazine, pubchem8621, acmc-1ca9d, ksc491g2h | |
| CCC1=NC=C(C)N=C1CC | |
| 150.23 | |
| 150.23 | |
| Liquid |
Safety and Handling
TSCA : Yes