missing translation for 'onlineSavingsMsg'
Learn More

2',3'-Dideoxyadenosine, 98%, Thermo Scientific Chemicals

Catalog No. AAJ64573MB
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ64573MB 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ64573MB Supplier Thermo Scientific Chemicals Supplier No. J64573MB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4097-22-7 | Molecular Formula: C10H13N5O2 | Molecular Weight: 235.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4097-22-7
Molecular Formula C10H13N5O2
Molecular Weight (g/mol) 235.25
MDL Number MFCD00010534
InChI Key WVXRAFOPTSTNLL-NKWVEPMBSA-N
Synonym 2',3'-dideoxyadenosine, dideoxyadenosine, adenosine, 2',3'-dideoxy, ddado, 2s,5r-5-6-amino-9h-purin-9-yl tetrahydrofuran-2-yl methanol, ccris 428, unii-4q86ah641a, d2a, dsstox_cid_3771, dsstox_rid_77189
PubChem CID 20039
IUPAC Name [(2S,5R)-5-(6-aminopurin-9-yl)oxolan-2-yl]methanol
SMILES NC1=C2N=CN([C@H]3CC[C@@H](CO)O3)C2=NC=N1

Specifications

Melting Point 181°C to 184°C
Color White
Quantity 25 mg
Beilstein 619924
Merck Index 14,3101
Formula Weight 235.24
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2',3'-Dideoxyadenosine
EINECSNumber 223-853-2
RTECSNumber AU7358900
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only