missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dicyanopyrazine, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 469000250

 View more versions of this product

Catalog No. AC469000250


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2,3-Dicyanopyrazine
131.0°C to 133.0°C
25 g
OTVZGAXESBAAQQ-UHFFFAOYSA-N
pyrazine-2,3-dicarbonitrile
130.11
13481-25-9
C6H2N4
Solubility aq. soln.: .. Other solubilities: soluble in methanol
N#CC1=NC=CN=C1C#N
130.11

Chemical Identifiers

13481-25-9
130.11
pyrazine-2,3-dicarbonitrile
C6H2N4
OTVZGAXESBAAQQ-UHFFFAOYSA-N
N#CC1=NC=CN=C1C#N
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement Harmful if swallowed.

Harmful in contact with skin.

Harmful if inhaled.

Causes skin irritation.

Causes serious eye irritation.

Harmful if inhaled.

GHS P Statement IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

Call a POISON CENTER or doctor/physician if you feel unwell.

IF ON SKIN: Wash with plenty of soap and water.

IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

IF IN EYES: Rinse cautiously with water for several minutes.

Remove contact lenses, if present and easy to do.

Continue rinsing.

Wear protective gloves/protective clothing/eye protection/face protection.

GHS Signal Word: Warning

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only