missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dicyanopyrazine, Thermo Scientific Chemicals

Catalog No. AC469000050
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC469000050 5 g
AC469000250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC469000050 Supplier Thermo Scientific Chemicals Supplier No. 469000050

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13481-25-9
Molecular Formula C6H2N4
Molecular Weight (g/mol) 130.11
InChI Key OTVZGAXESBAAQQ-UHFFFAOYSA-N
IUPAC Name pyrazine-2,3-dicarbonitrile
SMILES N#CC1=NC=CN=C1C#N

Specifications

Melting Point 131.0°C to 133.0°C
Quantity 5 g
Solubility Information Solubility aq. soln.: .. Other solubilities: soluble in methanol
Formula Weight 130.11
Chemical Name or Material 2,3-Dicyanopyrazine
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Harmful if swallowed.

Harmful in contact with skin.

Harmful if inhaled.

Causes skin irritation.

Causes serious eye irritation.

Harmful if inhaled.

Precautionary Statement GHS P Statement IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

Call a POISON CENTER or doctor/physician if you feel unwell.

IF ON SKIN: Wash with plenty of soap and water.

IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

IF IN EYES: Rinse cautiously with water for several minutes.

Remove contact lenses, if present and easy to do.

Continue rinsing.

Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only