missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Dicyanopyrazine 99.0+%, TCI America™
Supplier: TCI America D175125G
Specifications
| 2,3-Dicyanopyrazine | |
| 133°C | |
| MFCD00059771 | |
| Pyrazine-2,3-dicarbonitrile | |
| C1=CN=C(C(=N1)C#N)C#N | |
| 130.11 | |
| 130.11 | |
| Crystalline Powder |
| 13481-25-9 | |
| C6H2N4 | |
| 25 g | |
| OTVZGAXESBAAQQ-UHFFFAOYSA-N | |
| pyrazine-2,3-dicarbonitrile | |
| 300514 | |
| ≥99.0% (GC) |
Chemical Identifiers
| 13481-25-9 | |
| 130.11 | |
| OTVZGAXESBAAQQ-UHFFFAOYSA-N | |
| 300514 | |
| C1=CN=C(C(=N1)C#N)C#N |
| C6H2N4 | |
| MFCD00059771 | |
| Pyrazine-2,3-dicarbonitrile | |
| pyrazine-2,3-dicarbonitrile |
Safety and Handling
EINECSNumber : (9)-0533&(9)-1651
TSCA : No