missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Dicyano-5-methylpyrazine 99.0+%, TCI America™
$354.17 - $1243.32
Chemical Identifiers
| CAS | 52197-12-3 |
|---|---|
| Molecular Formula | C7H4N4 |
| Molecular Weight (g/mol) | 144.14 |
| MDL Number | MFCD00191635 |
| InChI Key | RHYUBLSWHDYKAO-UHFFFAOYSA-N |
| Synonym | 5-Methyl-2,3-pyrazinedicarbonitrile |
| PubChem CID | 300516 |
| IUPAC Name | 5-methylpyrazine-2,3-dicarbonitrile |
| SMILES | CC1=CN=C(C#N)C(=N1)C#N |
Chemical Identifiers
| 52197-12-3 | |
| 144.14 | |
| RHYUBLSWHDYKAO-UHFFFAOYSA-N | |
| 300516 | |
| CC1=CN=C(C#N)C(=N1)C#N |
| C7H4N4 | |
| MFCD00191635 | |
| 5-Methyl-2,3-pyrazinedicarbonitrile | |
| 5-methylpyrazine-2,3-dicarbonitrile |
Specifications
| 52197-12-3 | |
| White-Yellow | |
| MFCD00191635 | |
| 2811 | |
| RHYUBLSWHDYKAO-UHFFFAOYSA-N | |
| 5-methylpyrazine-2,3-dicarbonitrile | |
| 300516 | |
| ≥99.0% (GC) | |
| 2,3-Dicyano-5-methylpyrazine |
| 100°C | |
| C7H4N4 | |
| 5 g | |
| 5-Methyl-2,3-pyrazinedicarbonitrile | |
| CC1=CN=C(C#N)C(=N1)C#N | |
| 144.14 | |
| 144.14 | |
| Crystalline Powder |
Safety and Handling
TSCA : No