missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichlorotoluene 98.0+%, TCI America™

Catalog No. D1805500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1805500G 500 g
D180525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D1805500G Supplier TCI America Supplier No. D1805500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 32768-54-0
Molecular Formula C7H6Cl2
Molecular Weight (g/mol) 161.025
MDL Number MFCD00000546
InChI Key GWLKCPXYBLCEKC-UHFFFAOYSA-N
Synonym 2,3-dichlorotoluene, benzene, 1,2-dichloro-3-methyl, toluene, 2,3-dichloro, 1,2-dichloro-3-methyl-benzene, acmc-209hvi, ksc490m7b, gwlkcpxyblcekc-uhfffaoysa, 2,3-dct, #, labotest-bb ltbb004413
PubChem CID 34702
IUPAC Name 1,2-dichloro-3-methylbenzene
SMILES CC1=C(C(=CC=C1)Cl)Cl

Specifications

Color Colorless
Boiling Point 208°C
Quantity 500 g
Formula Weight 161.03
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,3-Dichlorotoluene
EINECSNumber (3)-0078
TSCA Yes