missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Dichloroquinoxaline 98.0+%, TCI America™
Supplier: TCI America D262225G
Specifications
| 2,3-Dichloroquinoxaline | |
| 153°C | |
| MFCD00006720 | |
| 2811 | |
| SPSSDDOTEZKOOV-UHFFFAOYSA-N | |
| 2,3-dichloroquinoxaline | |
| 16659 | |
| ≥98.0% (GC) |
| 2213-63-0 | |
| C8H4Cl2N2 | |
| 25 g | |
| quinoxaline, 2,3-dichloro, 2,3-dichloro-quinoxaline, ccris 4789, quinoxaline, dichloro, dichloroquinoxaline, pubchem15473, 2,3 dichloroquinoxaline, 2.3-dichloroquinoxaline, acmc-1cfck, 3,4-dichloroquinoxaline | |
| C1=CC=C2C(=C1)N=C(C(=N2)Cl)Cl | |
| 199.034 | |
| 199.03 | |
| Crystalline Powder |
Chemical Identifiers
| 2213-63-0 | |
| 199.034 | |
| SPSSDDOTEZKOOV-UHFFFAOYSA-N | |
| 16659 | |
| C1=CC=C2C(=C1)N=C(C(=N2)Cl)Cl |
| C8H4Cl2N2 | |
| MFCD00006720 | |
| quinoxaline, 2,3-dichloro, 2,3-dichloro-quinoxaline, ccris 4789, quinoxaline, dichloro, dichloroquinoxaline, pubchem15473, 2,3 dichloroquinoxaline, 2.3-dichloroquinoxaline, acmc-1cfck, 3,4-dichloroquinoxaline | |
| 2,3-dichloroquinoxaline |
Safety and Handling
RTECSNumber : VD1720000
TSCA : Yes