missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichloroquinoxaline, 98%

CAS: 2213-63-0 | C8H4Cl2N2 | 199.034 g/mol

$293.04 - $293.04

Chemical Identifiers

CAS 2213-63-0
Molecular Formula C8H4Cl2N2
Molecular Weight (g/mol) 199.034
MDL Number MFCD00006720
InChI Key SPSSDDOTEZKOOV-UHFFFAOYSA-N
Synonym quinoxaline, 2,3-dichloro, 2,3-dichloro-quinoxaline, ccris 4789, quinoxaline, dichloro, dichloroquinoxaline, pubchem15473, 2,3 dichloroquinoxaline, 2.3-dichloroquinoxaline, acmc-1cfck, 3,4-dichloroquinoxaline
PubChem CID 16659
IUPAC Name 2,3-dichloroquinoxaline
SMILES C1=CC=C2C(=C1)N=C(C(=N2)Cl)Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1173414
View Documents
Thermo Scientific Chemicals
A1173414
25 g
Each for $293.04
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2213-63-0
199.034
SPSSDDOTEZKOOV-UHFFFAOYSA-N
16659
C1=CC=C2C(=C1)N=C(C(=N2)Cl)Cl
C8H4Cl2N2
MFCD00006720
quinoxaline, 2,3-dichloro, 2,3-dichloro-quinoxaline, ccris 4789, quinoxaline, dichloro, dichloroquinoxaline, pubchem15473, 2,3 dichloroquinoxaline, 2.3-dichloroquinoxaline, acmc-1cfck, 3,4-dichloroquinoxaline
2,3-dichloroquinoxaline

Specifications

2213-63-0
Odorless
MFCD00006720
UN2811
quinoxaline, 2,3-dichloro, 2,3-dichloro-quinoxaline, ccris 4789, quinoxaline, dichloro, dichloroquinoxaline, pubchem15473, 2,3 dichloroquinoxaline, 2.3-dichloroquinoxaline, acmc-1cfck, 3,4-dichloroquinoxaline
C1=CC=C2C(=C1)N=C(C(=N2)Cl)Cl
199.034
199.04
2,3-Dichloroquinoxaline
151°C to 155°C
C8H4Cl2N2
25 g
126076
SPSSDDOTEZKOOV-UHFFFAOYSA-N
2,3-dichloroquinoxaline
16659
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H301-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 218-667-3

RTECSNumber : VD1720000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only