missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Dichloropyridine-4-boronic Acid (contains varying amounts of Anhydride), TCI America™
$152.86 - $152.86
Specifications
| CAS | 951677-39-7 |
|---|---|
| Melting Point | 141°C |
| Color | White |
| Molecular Formula | C5H4BCl2NO2 |
| MDL Number | MFCD04038756 |
Specifications
| 951677-39-7 | |
| White | |
| MFCD04038756 | |
| (2,3-dichloropyridin-4-yl)boronic acid | |
| 191.80 |
| 141°C | |
| C5H4BCl2NO2 | |
| ABRTZUPJNVJMHP-UHFFFAOYSA-N | |
| 3478955 | |
| Crystalline Powder |