missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichloropropionamide 98.0+%, TCI America™

Catalog No. D199910G
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D199910G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D199910G Supplier TCI America Supplier No. D199910G

Chemical Identifiers

CAS 19433-84-2
Molecular Formula C3H5Cl2NO
Molecular Weight (g/mol) 141.979
MDL Number MFCD00040311
InChI Key SQGUQSRDWXDLIH-UHFFFAOYSA-N
PubChem CID 86874
IUPAC Name 2,3-dichloropropanamide
SMILES C(C(C(=O)N)Cl)Cl

Specifications

Melting Point 106°C
Color White-Yellow
Quantity 10 g
Formula Weight 141.98
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Dichloropropionamide