missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Dichlorophenylboronic Acid (contains varying amounts of Anhydride), TCI America™
$117.43 - $117.43
Chemical Identifiers
| CAS | 151169-74-3 |
|---|---|
| Molecular Formula | C6H5BCl2O2 |
| Molecular Weight (g/mol) | 190.81 |
| MDL Number | MFCD01075703 |
| InChI Key | TYIKXPOMOYDGCS-UHFFFAOYSA-N |
| PubChem CID | 2734661 |
| IUPAC Name | (2,3-dichlorophenyl)boronic acid |
| SMILES | OB(O)C1=C(Cl)C(Cl)=CC=C1 |
Chemical Identifiers
| 151169-74-3 | |
| 190.81 | |
| TYIKXPOMOYDGCS-UHFFFAOYSA-N | |
| (2,3-dichlorophenyl)boronic acid |
| C6H5BCl2O2 | |
| MFCD01075703 | |
| 2734661 | |
| OB(O)C1=C(Cl)C(Cl)=CC=C1 |
Specifications
| 151169-74-3 | |
| C6H5BCl2O2 | |
| 5 g | |
| TYIKXPOMOYDGCS-UHFFFAOYSA-N | |
| (2,3-dichlorophenyl)boronic acid | |
| 2734661 | |
| Crystalline Powder |
| 125°C | |
| MFCD01075703 | |
| 2,3-dichlorophenyl boronic acid, 2,3-dichlorobenzeneboronic acid, 2,3-dichlorophenyl boranediol, boronic acid, 2,3-dichlorophenyl, contains varying amounts of anhydride, pubchem1810, dichlorophenylboronic acid, acmc-1c0ur, dichlorobenzene boronic acid | |
| OB(O)C1=C(Cl)C(Cl)=CC=C1 | |
| 190.81 | |
| 190.81 | |
| 2,3-Dichlorophenylboronic Acid (contains varying amounts of Anhydride) |
Safety and Handling
TSCA : No