missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichlorophenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. D34345G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D34345G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D34345G Supplier TCI America Supplier No. D34345G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 151169-74-3
Molecular Formula C6H5BCl2O2
Molecular Weight (g/mol) 190.81
MDL Number MFCD01075703
InChI Key TYIKXPOMOYDGCS-UHFFFAOYSA-N
PubChem CID 2734661
IUPAC Name (2,3-dichlorophenyl)boronic acid
SMILES OB(O)C1=C(Cl)C(Cl)=CC=C1

Specifications

Melting Point 125°C
Quantity 5 g
Synonym 2,3-dichlorophenyl boronic acid, 2,3-dichlorobenzeneboronic acid, 2,3-dichlorophenyl boranediol, boronic acid, 2,3-dichlorophenyl, contains varying amounts of anhydride, pubchem1810, dichlorophenylboronic acid, acmc-1c0ur, dichlorobenzene boronic acid
Formula Weight 190.81
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Dichlorophenylboronic Acid (contains varying amounts of Anhydride)