missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichlorophenylacetic Acid 98.0+%, TCI America™

Catalog No. d29315g
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D29315G 5 g
D293125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D29315G Supplier TCI America Supplier No. D29315G
Only null left

Chemical Identifiers

CAS 10236-60-9
Molecular Formula C8H6Cl2O2
Molecular Weight (g/mol) 205.034
MDL Number MFCD01861393
InChI Key YWMXEUIQZOQESD-UHFFFAOYSA-N
Synonym 2,3-dichlorophenylacetic acid, 2-2,3-dichlorophenyl acetic acid, benzeneacetic acid, 2,3-dichloro, 2,3-dichlorophenyl acetic acid, 2,3-dichlorobenzeneacetic acid, rarechem al bo 1228, timtec-bb sbb003500, 3-dichlorophenylacetic acid, acmc-1c25x, 2,3-dichlorophenylaceticacid
PubChem CID 2734600
IUPAC Name 2-(2,3-dichlorophenyl)acetic acid
SMILES C1=CC(=C(C(=C1)Cl)Cl)CC(=O)O

Specifications

Melting Point 134°C
Color White
Quantity 5 g
Formula Weight 205.03
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Dichlorophenylacetic Acid