missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichlorophenylacetic acid, 98%

Catalog No. AAH3276306
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3276306 5 g
AAH3276314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3276306 Supplier Thermo Scientific Chemicals Supplier No. H3276306

CAS: 10236-60-9 | C8H6Cl2O2 | 205.034 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10236-60-9
Molecular Formula C8H6Cl2O2
Molecular Weight (g/mol) 205.034
MDL Number MFCD01861393
InChI Key YWMXEUIQZOQESD-UHFFFAOYSA-N
Synonym 2,3-dichlorophenylacetic acid, 2-2,3-dichlorophenyl acetic acid, benzeneacetic acid, 2,3-dichloro, 2,3-dichlorophenyl acetic acid, 2,3-dichlorobenzeneacetic acid, rarechem al bo 1228, timtec-bb sbb003500, 3-dichlorophenylacetic acid, acmc-1c25x, 2,3-dichlorophenylaceticacid
PubChem CID 2734600
IUPAC Name 2-(2,3-dichlorophenyl)acetic acid
SMILES C1=CC(=C(C(=C1)Cl)Cl)CC(=O)O

Specifications

Melting Point 132°C to 136°C
Odor Odorless
Quantity 5 g
Formula Weight 205.04
Percent Purity 98%
Chemical Name or Material 2,3-Dichlorophenylacetic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only