missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Dichloromaleic anhydride, 97%, Thermo Scientific™
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
| Infrared Spectrum | Authentic |
| Assay Percent Range | 96% min. (GC) |
| Quantity | 100g |
| Beilstein | 17,434 |
| Fieser | 04,135; 05,195 |
| Formula Weight | 166.95 |
| Percent Purity | 97% |
| Chemical Name or Material | 2,3-Dichloromaleic anhydride |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician if you feel unwell. |
| EINECSNumber | 214-343- |
RUO – Research Use Only