missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Dichloroisobutyric Acid 98.0+%, TCI America™
Specifications
Specifications
| Color | Colorless |
| Boiling Point | 113°C |
| Quantity | 10 g |
| UN Number | 3265 |
| Formula Weight | 156.99 |
| Percent Purity | ≥98.0% (T) |
| Physical Form | Liquid |
| Chemical Name or Material | 2,3-Dichloroisobutyric Acid |
Safety and Handling
| TSCA | No |