missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichlorobenzeneboronic acid, 98%

Catalog No. p-7026664
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2278103 1 g
AAB2278106 5 g
AAB2278114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2278103 Supplier Thermo Scientific Chemicals Supplier No. B2278103
Only null left

CAS: 151169-74-3 | C6H5BCl2O2 | 190.81 g/mol

2,3-Dichlorobenzeneboronic acid is used as a reactant for Suzuki-Miyaura coupling reactions. It is also used in preparation of biologically and pharmacologically active molecules.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3-Dichlorobenzeneboronic acid is used as a reactant for Suzuki-Miyaura coupling reactions. It is also used in preparation of biologically and pharmacologically active molecules.

Solubility
Soluble in methanol.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 151169-74-3
Molecular Formula C6H5BCl2O2
Molecular Weight (g/mol) 190.81
MDL Number MFCD01075703
InChI Key TYIKXPOMOYDGCS-UHFFFAOYSA-N
Synonym 2,3-dichlorophenyl boronic acid, 2,3-dichlorobenzeneboronic acid, 2,3-dichlorophenyl boranediol, boronic acid, 2,3-dichlorophenyl, contains varying amounts of anhydride, pubchem1810, dichlorophenylboronic acid, acmc-1c0ur, dichlorobenzene boronic acid
PubChem CID 2734661
IUPAC Name (2,3-dichlorophenyl)boronic acid
SMILES OB(O)C1=C(Cl)C(Cl)=CC=C1

Specifications

Melting Point 233°C to 242°C
Quantity 1 g
Solubility Information Soluble in methanol.
Formula Weight 190.82
Percent Purity 98%
Chemical Name or Material 2,3-Dichlorobenzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only