missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichlorobenzaldehyde 98.0+%, TCI America™

Catalog No. D1666500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1666500G 500 g
D166625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D1666500G Supplier TCI America Supplier No. D1666500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6334-18-5
Molecular Formula C7H4Cl2O
Molecular Weight (g/mol) 175.01
MDL Number MFCD00010127
InChI Key LLMLNAVBOAMOEE-UHFFFAOYSA-N
Synonym benzaldehyde, 2,3-dichloro, dichlorobenzaldehyde, benzaldehyde, dichloro, ccris 8612, dichlorbenzaldehyd, pubchem3079, 2,3-dichlorobenzaldehyd, benzaldehyde,3-dichloro, 2,3-dichloro-benzaldehyde, acmc-209nf0
PubChem CID 35745
IUPAC Name 2,3-dichlorobenzaldehyde
SMILES ClC1=CC=CC(C=O)=C1Cl

Specifications

Melting Point 62°C
Color White-Yellow
Boiling Point 105°C
Quantity 500 g
UN Number 1759
Formula Weight 175.01
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Dichlorobenzaldehyde
EINECSNumber (3)-4144
TSCA Yes