missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichloro-5-hydroxypyridine 98.0+%, TCI America™

Catalog No. D48991G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D48991G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D48991G Supplier TCI America Supplier No. D48991G

Chemical Identifiers

CAS 110860-92-9
Molecular Formula C5H3Cl2NO
Molecular Weight (g/mol) 163.985
MDL Number MFCD09835219
InChI Key GMHPKIGFTDRQBP-UHFFFAOYSA-N
Synonym 5,6-Dichloro-3-pyridinol
PubChem CID 14391672
IUPAC Name 5,6-dichloropyridin-3-ol
SMILES C1=C(C=NC(=C1Cl)Cl)O

Specifications

Melting Point 186°C
Quantity 1 g
Formula Weight 163.99
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Dichloro-5-hydroxypyridine