Learn More
2,3-Dichloro-1-propene, 98%
CAS: 78-88-6 | C3H4Cl2 | 110.965 g/mol
Supplier: Thermo Scientific Chemicals L0894318
Description
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Soluble in water (2g/L).
Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications
| 2,3-Dichloro-1-propene | |
| -83°C | |
| 93°C to 96°C | |
| Pungent | |
| 1.462 | |
| MFCD00000943 | |
| UN2047 | |
| 2,3-dichloropropene, 2,3-dichloro-1-propene, 2-chloroallyl chloride, 2,3-dichloropropylene, 1-propene, 2,3-dichloro, propene, 2,3-dichloro, 1,2-dichloro-2-propene, propylene, 2,3-dichloro, ccris 956, unii-370tnk5i41 | |
| FALCMQXTWHPRIH-UHFFFAOYSA-N | |
| 2,3-dichloroprop-1-ene | |
| 6565 | |
| 98% |
| 78-88-6 | |
| 1.209 | |
| 10°C (50°F) | |
| C3H4Cl2 | |
| ClCH2CCl=CH2 | |
| 50 g | |
| 1361491 | |
| Soluble in water (2g/L). | |
| C=C(CCl)Cl | |
| 110.965 | |
| 110.97 |
Chemical Identifiers
| 78-88-6 | |
| 110.965 | |
| FALCMQXTWHPRIH-UHFFFAOYSA-N | |
| 6565 | |
| C=C(CCl)Cl |
| C3H4Cl2 | |
| MFCD00000943 | |
| 2,3-dichloropropene, 2,3-dichloro-1-propene, 2-chloroallyl chloride, 2,3-dichloropropylene, 1-propene, 2,3-dichloro, propene, 2,3-dichloro, 1,2-dichloro-2-propene, propylene, 2,3-dichloro, ccris 956, unii-370tnk5i41 | |
| 2,3-dichloroprop-1-ene |
Safety and Handling
GHS H Statement
H225-H318-H341-H302-H312-H332-H335-H315
Highly flammable liquid and vapor.
Causes serious eye damage.
Suspected of causing genetic defects.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
May cause respiratory irritation.
Causes skin irritation.
P201-P202-P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P281-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P310-P312-P330-P332+P313-P363-P370+P378q-P50
H225-H302+H312+H332-H315-H318-H335-H340
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: DICHLOROPROPENES
EINECSNumber : 201-153-8
RTECSNumber : UC8400000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only