missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichloro-1-propene, 98%

Catalog No. AAL0894330
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0894330 250 g
AAL0894318 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0894330 Supplier Thermo Scientific Chemicals Supplier No. L0894330
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 78-88-6 | C3H4Cl2 | 110.965 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in water (2g/L).

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 78-88-6
Molecular Formula C3H4Cl2
Molecular Weight (g/mol) 110.965
MDL Number MFCD00000943
InChI Key FALCMQXTWHPRIH-UHFFFAOYSA-N
Synonym 2,3-dichloropropene, 2,3-dichloro-1-propene, 2-chloroallyl chloride, 2,3-dichloropropylene, 1-propene, 2,3-dichloro, propene, 2,3-dichloro, 1,2-dichloro-2-propene, propylene, 2,3-dichloro, ccris 956, unii-370tnk5i41
PubChem CID 6565
IUPAC Name 2,3-dichloroprop-1-ene
SMILES C=C(CCl)Cl

Specifications

Melting Point -83°C
Density 1.209
Boiling Point 93°C to 96°C
Flash Point 10°C (50°F)
Odor Pungent
Refractive Index 1.462
Linear Formula ClCH2CCl=CH2
Quantity 250 g
UN Number UN2047
Beilstein 1361491
Solubility Information Soluble in water (2g/L).
Formula Weight 110.97
Percent Purity 98%
Chemical Name or Material 2,3-Dichloro-1-propene
Show More Show Less
Hazard Category H225-H302+H312+H332-H315-H318-H335-H340
Hazard Statement GHS H Statement
H225-H318-H341-H302-H312-H332-H335-H315
Highly flammable liquid and vapor.
Causes serious eye damage.
Suspected of causing genetic defects.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement P201-P202-P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P281-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P310-P312-P330-P332+P313-P363-P370+P378q-P50
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: DICHLOROPROPENES
EINECSNumber 201-153-8
RTECSNumber UC8400000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only