missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichloro-1-propanol 98.0+%, TCI America™

Catalog No. d040325g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
25 g
500 g
Catalog No. Quantity
D040325G 25 g
D0403100G 100 g
D0403500G 500 g
3 options

Catalog No. D040325G

Supplier: TCI America D040325G

Only null left

Chemical Identifiers

CAS 616-23-9
Molecular Formula C3H6Cl2O
Molecular Weight (g/mol) 128.98
MDL Number MFCD00040446
InChI Key ZXCYIJGIGSDJQQ-UHFFFAOYSA-N
Synonym 2,3-dichloro-1-propanol, 1-propanol, 2,3-dichloro, 2,3-dichloropropanol, alpha,beta-dichlorohydrin, 1,2-dichloro-3-propanol, glycerol alpha,beta-dichlorohydrin, 2,3-dichloropropyl alcohol, glycerol 1,2-dichlorohydrin, 2,3-dichloropropane-1-ol, ccris 954
PubChem CID 12018
IUPAC Name 2,3-dichloropropan-1-ol
SMILES C(C(CCl)Cl)O

Specifications

Color Colorless
Boiling Point 182°C
Quantity 25 g
UN Number 2810
Formula Weight 128.98
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,3-Dichloro-1-propanol
EINECSNumber (2)-2002
RTECSNumber UB1225000
TSCA Yes