missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Dichloro-1-propanol 98.0+%, TCI America™
Supplier: TCI America D0403100G
Specifications
| 2,3-Dichloro-1-propanol | |
| Colorless | |
| C3H6Cl2O | |
| 100 g | |
| 2,3-dichloro-1-propanol, 1-propanol, 2,3-dichloro, 2,3-dichloropropanol, alpha,beta-dichlorohydrin, 1,2-dichloro-3-propanol, glycerol alpha,beta-dichlorohydrin, 2,3-dichloropropyl alcohol, glycerol 1,2-dichlorohydrin, 2,3-dichloropropane-1-ol, ccris 954 | |
| C(C(CCl)Cl)O | |
| 128.98 | |
| 128.98 | |
| Liquid |
| 616-23-9 | |
| 182°C | |
| MFCD00040446 | |
| 2810 | |
| ZXCYIJGIGSDJQQ-UHFFFAOYSA-N | |
| 2,3-dichloropropan-1-ol | |
| 12018 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 616-23-9 | |
| 128.98 | |
| ZXCYIJGIGSDJQQ-UHFFFAOYSA-N | |
| 12018 | |
| C(C(CCl)Cl)O |
| C3H6Cl2O | |
| MFCD00040446 | |
| 2,3-dichloro-1-propanol, 1-propanol, 2,3-dichloro, 2,3-dichloropropanol, alpha,beta-dichlorohydrin, 1,2-dichloro-3-propanol, glycerol alpha,beta-dichlorohydrin, 2,3-dichloropropyl alcohol, glycerol 1,2-dichlorohydrin, 2,3-dichloropropane-1-ol, ccris 954 | |
| 2,3-dichloropropan-1-ol |
Safety and Handling
EINECSNumber : (2)-2002
RTECSNumber : UB1225000
TSCA : Yes