missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichloro-1-propanol, 97+%

Catalog No. AAA1844406
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1844406 5 g
AAA1844414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1844406 Supplier Thermo Scientific Chemicals Supplier No. A1844406
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 616-23-9 | C3H6Cl2O | 128.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 616-23-9
Molecular Formula C3H6Cl2O
Molecular Weight (g/mol) 128.98
MDL Number MFCD00040446
InChI Key ZXCYIJGIGSDJQQ-UHFFFAOYSA-N
Synonym 2,3-dichloro-1-propanol, 1-propanol, 2,3-dichloro, 2,3-dichloropropanol, alpha,beta-dichlorohydrin, 1,2-dichloro-3-propanol, glycerol alpha,beta-dichlorohydrin, 2,3-dichloropropyl alcohol, glycerol 1,2-dichlorohydrin, 2,3-dichloropropane-1-ol, ccris 954
PubChem CID 12018
IUPAC Name 2,3-dichloropropan-1-ol
SMILES C(C(CCl)Cl)O

Specifications

Density 1.361
Boiling Point 182°C
Flash Point 93°C (199°F)
Refractive Index 1.485
Quantity 5 g
UN Number UN2810
Beilstein 1732060
Formula Weight 128.99
Percent Purity ≥97%
Chemical Name or Material 2,3-Dichloro-1-propanol
Hazard Category H227-H301-H310-H315-H318-H332-H341
Hazard Statement GHS H Statement
H301-H310-H318-H341-H227-H315-H335
Toxic if swallowed.
Fatal in contact with skin.
Causes serious eye damage.
Suspected of causing genetic defects.
Combustible liquid.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P210-P261-P262-P264b-P270-P271-P281-P301+P310-P302+P350-P304+P340-P305+P351+P338-P308+P313-P310-P330-P332+P313-P361-P363-P370+P378q-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: II
EINECSNumber 210-470-0
RTECSNumber UB1225000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only