missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dibromothiophene 95.0+%, TCI America™

Catalog No. D15485G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D15485G 5 g
D154825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D15485G Supplier TCI America Supplier No. D15485G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3140-93-0
Molecular Formula C4H2Br2S
Molecular Weight (g/mol) 241.928
MDL Number MFCD00005418
InChI Key ATRJNSFQBYKFSM-UHFFFAOYSA-N
Synonym thiophene, 2,3-dibromo, 2,3-dibromo thiophene, dibromothiophene, 2,3-dibromo-thiophene, 2,3-dibromthiophen, 2 3-dibromothiophene, pubchem5511, thiophene,3-dibromo, acmc-1ckid, ksc491i5b
PubChem CID 76590
IUPAC Name 2,3-dibromothiophene
SMILES C1=CSC(=C1Br)Br

Specifications

Color Yellow
Boiling Point 219°C
Quantity 5 g
UN Number 1993
Formula Weight 241.93
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,3-Dibromothiophene