missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dibromothiophene, 98+%

Catalog No. AAA1299814
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1299814 25 g
AAA1299806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1299814 Supplier Thermo Scientific Chemicals Supplier No. A1299814
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3140-93-0 | C4H2Br2S | 241.928 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3140-93-0
Molecular Formula C4H2Br2S
Molecular Weight (g/mol) 241.928
MDL Number MFCD00005418
InChI Key ATRJNSFQBYKFSM-UHFFFAOYSA-N
Synonym thiophene, 2,3-dibromo, 2,3-dibromo thiophene, dibromothiophene, 2,3-dibromo-thiophene, 2,3-dibromthiophen, 2 3-dibromothiophene, pubchem5511, thiophene,3-dibromo, acmc-1ckid, ksc491i5b
PubChem CID 76590
IUPAC Name 2,3-dibromothiophene
SMILES C1=CSC(=C1Br)Br

Specifications

Density 2.135
Boiling Point 215°C to 216°C
Flash Point >100°C (212°F)
Refractive Index 1.631
Quantity 25 g
Beilstein 107512
Sensitivity Light sensitive
Formula Weight 241.94
Percent Purity ≥98%
Chemical Name or Material 2,3-Dibromothiophene
Hazard Category H226-H301-H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Hazard Class: 3; Packaging Group: III
EINECSNumber 221-542-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only