missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dibromopyridine 98.0+%, TCI America™

Catalog No. D29175G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D29175G 5 g
D29171G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D29175G Supplier TCI America Supplier No. D29175G

Chemical Identifiers

CAS 13534-89-9
Molecular Formula C5H3Br2N
Molecular Weight (g/mol) 236.894
MDL Number MFCD00234014
InChI Key SLMHHOVQRSSRCV-UHFFFAOYSA-N
Synonym pyridine, 2,3-dibromo, dibromopyridine, 2,3-dibromo pyridine, pubchem5402, acmc-209c0b, 2,3-dibromopyridine, ksc492s9r, tpc-py084, slmhhovqrssrcv-uhfffaoysa, abbypharma ap-12-5128
PubChem CID 817102
IUPAC Name 2,3-dibromopyridine
SMILES C1=CC(=C(N=C1)Br)Br

Specifications

Melting Point 59°C
Boiling Point 249°C
Quantity 5 g
Formula Weight 236.89
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Dibromopyridine