missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dibromopyridine, 97%

Catalog No. AAL1952706
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1952706 5 g
AAL1952714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1952706 Supplier Thermo Scientific Chemicals Supplier No. L1952706

CAS: 13534-89-9 | C5H3Br2N | 236.894 g/mol

It is employed as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Moisture sensitive. Store in dry condition. Keep oxidizing agents.

Chemical Identifiers

CAS 13534-89-9
Molecular Formula C5H3Br2N
Molecular Weight (g/mol) 236.894
MDL Number MFCD00234014
InChI Key SLMHHOVQRSSRCV-UHFFFAOYSA-N
Synonym pyridine, 2,3-dibromo, dibromopyridine, 2,3-dibromo pyridine, pubchem5402, acmc-209c0b, 2,3-dibromopyridine, ksc492s9r, tpc-py084, slmhhovqrssrcv-uhfffaoysa, abbypharma ap-12-5128
PubChem CID 817102
IUPAC Name 2,3-dibromopyridine
SMILES C1=CC(=C(N=C1)Br)Br

Specifications

Melting Point 58°C to 60°C
Boiling Point 249°C to 250°C
Quantity 5 g
Beilstein 109828
Solubility Information Insoluble in water.
Formula Weight 236.9
Percent Purity 97%
Chemical Name or Material 2,3-Dibromopyridine
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only