missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dibromopyridine-4-carboxylic acid, 97%, Thermo Scientific Chemicals

$259.49 - $259.49

Chemical Identifiers

CAS 1020056-98-7
Molecular Formula C6H3Br2NO2
Molecular Weight (g/mol) 280.903
MDL Number MFCD09997708
InChI Key ICISUZSRQHUJKX-UHFFFAOYSA-N
Synonym 2,3-dibromoisonicotinic acid, 2,3-dibromo-4-pyridinecarboxylic acid, acmc-20ahus, 2,3-dibromopyridine-4-carboxylicacid
PubChem CID 26967070
IUPAC Name 2,3-dibromopyridine-4-carboxylic acid
SMILES C1=CN=C(C(=C1C(=O)O)Br)Br
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH3170803
View Documents
Thermo Scientific Chemicals
H3170803
1 g
Each for $259.49
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1020056-98-7
280.903
ICISUZSRQHUJKX-UHFFFAOYSA-N
26967070
C1=CN=C(C(=C1C(=O)O)Br)Br
C6H3Br2NO2
MFCD09997708
2,3-dibromoisonicotinic acid, 2,3-dibromo-4-pyridinecarboxylic acid, acmc-20ahus, 2,3-dibromopyridine-4-carboxylicacid
2,3-dibromopyridine-4-carboxylic acid

Specifications

1020056-98-7
MFCD09997708
2,3-dibromoisonicotinic acid, 2,3-dibromo-4-pyridinecarboxylic acid, acmc-20ahus, 2,3-dibromopyridine-4-carboxylicacid
C1=CN=C(C(=C1C(=O)O)Br)Br
280.903
280.9
2,3-Dibromopyridine-4-carboxylic acid
C6H3Br2NO2
1 g
ICISUZSRQHUJKX-UHFFFAOYSA-N
2,3-dibromopyridine-4-carboxylic acid
26967070
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only