missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dibromopropionyl chloride, 95%

Catalog No. p-7023803
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1771209 10 g
AAA1771218 50 g
AAA1771230 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1771209 Supplier Thermo Scientific Chemicals Supplier No. A1771209
Only null left

CAS: 18791-02-1 | C3H3Br2ClO | 250.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 18791-02-1
Molecular Formula C3H3Br2ClO
Molecular Weight (g/mol) 250.31
MDL Number MFCD00000712
InChI Key HWKWYDXHMQQDQJ-UHFFFAOYNA-N
PubChem CID 86790
IUPAC Name 2,3-dibromopropanoyl chloride
SMILES ClC(=O)C(Br)CBr

Specifications

Density 2.181
Boiling Point 191°C to 193°C
Flash Point 67°C (152.6°F)
Odor Acrid
Refractive Index 1.542
Quantity 10 g
UN Number UN3265
Beilstein 1749801
Sensitivity Moisture sensitive
Synonym 2,3-dibromopropionyl chloride, propanoyl chloride, 2,3-dibromo, alpha,beta-dibromopropionyl chloride, propanoyl chloride,2,3-dibromo, propionyl chloride, 2,3-dibromo, acmc-209eqo, propanoyl chloride, 2,3-dibromo-9ci, 2,3-dibromopropanyl chloride, 3-02-00-00572 beilstein handbook reference
Formula Weight 250.32
Percent Purity 95%
Chemical Name or Material 2,3-Dibromopropionyl chloride
Show More Show Less
Hazard Category H227-H290-H302-H314-H335
Hazard Statement GHS H Statement
H301-H314-H318-H227-H332
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Harmful if inhaled.
Precautionary Statement P210-P234-P235-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P390-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 242-575-2
RTECSNumber UG6657500
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only