missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dibromobutyric acid, 97%

Catalog No. p-7044143
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0484106 5 g
AAL0484114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0484106 Supplier Thermo Scientific Chemicals Supplier No. L0484106

CAS: 600-30-6 | C4H6Br2O2 | 245.898 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 600-30-6
Molecular Formula C4H6Br2O2
Molecular Weight (g/mol) 245.898
MDL Number MFCD00014334
InChI Key HESQKTULJLBDRF-UHFFFAOYSA-N
Synonym 2,3-dibromobutyric acid, .alpha.,.beta.-dibromobutyric acid, 2,3-dibromo-butyric acid, acmc-1atov, 2,3 dibromobutyric acid, #, butanoic acid,2,3-dibromo, butanoic acid, 2,3-dibromo, alpha,beta-dibromo-n-butyric acid, .alpha.,.beta.-dibromo-n-butyric acid
PubChem CID 95386
IUPAC Name 2,3-dibromobutanoic acid
SMILES CC(C(C(=O)O)Br)Br

Specifications

Melting Point 84°C to 88°C
Boiling Point 100°C to 110°C (2 mmHg)
Quantity 5 g
UN Number UN3261
Beilstein 1721669
Formula Weight 245.91
Percent Purity 97%
Chemical Name or Material 2,3-Dibromobutyric acid
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 209-992-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only