missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dibromo-1-propanol 95.0+%, TCI America™

Catalog No. D020425G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D020425G 25 g
D0204500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D020425G Supplier TCI America Supplier No. D020425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 96-13-9
Molecular Formula C3H6Br2O
Molecular Weight (g/mol) 217.89
MDL Number MFCD00004699
InChI Key QWVCIORZLNBIIC-UHFFFAOYNA-N
Synonym 2,3-dibromo-1-propanol, 2,3-dibromopropanol, 1-propanol, 2,3-dibromo, dibromopropanol, brominex 257, 2,3-dibromopropyl alcohol, 1,2-dibromopropan-3-ol, usaf do-42, dbp flame retardant, beta-dibromohydrin
PubChem CID 7281
ChEBI CHEBI:19310
IUPAC Name 2,3-dibromopropan-1-ol
SMILES OCC(Br)CBr

Specifications

Melting Point 9°C
Color Yellow
Boiling Point 99°C
Quantity 25 g
UN Number 2810
Formula Weight 217.89
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,3-Dibromo-1-propanol
EINECSNumber (2)-2002
RTECSNumber UB0175000
TSCA Yes