missing translation for 'onlineSavingsMsg'
Learn More

2,3-Diaminotoluene, 97%

Catalog No. AAA1564706
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1564706 5 g
AAA1564714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1564706 Supplier Thermo Scientific Chemicals Supplier No. A1564706
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2687-25-4 | C7H10N2 | 122.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2687-25-4
Molecular Formula C7H10N2
Molecular Weight (g/mol) 122.17
MDL Number MFCD00011589
InChI Key AXNUJYHFQHQZBE-UHFFFAOYSA-N
Synonym 2,3-diaminotoluene, toluene-2,3-diamine, 3-methyl-o-phenylenediamine, 1,2-benzenediamine, 3-methyl, 3-methyl-1,2-phenylenediamine, 2,3-tolylenediamine, 2,3-toluylenediamine, 1,2-diamino-3-methylbenzene, 3-methyl-1,2-benzenediamine, 1-methyl-2,3-phenylenediamine
PubChem CID 17593
IUPAC Name 3-methylbenzene-1,2-diamine
SMILES CC1=CC=CC(N)=C1N

Specifications

Melting Point 56°C to 60°C
Boiling Point 225°C
Quantity 5 g
Formula Weight 122.17
Percent Purity 98%
Chemical Name or Material 2,3-Diaminotoluene
Hazard Category H301-H312+H332-H317-H319-H341-H350-H361f-H373
Hazard Statement GHS H Statement
H301-H312-H315-H319-H335
Toxic if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P272-P280-P281-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P330-P333+P313-P363-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 220-248-5
RTECSNumber XS9550000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only