missing translation for 'onlineSavingsMsg'
Learn More

2,3-Diaminotoluene, 97%, Thermo Scientific™

$62.32 - $198.52

Chemical Identifiers

CAS 2687-25-4
Molecular Formula C7H10N2
Molecular Weight (g/mol) 122.17
MDL Number MFCD00011589
InChI Key AXNUJYHFQHQZBE-UHFFFAOYSA-N
Synonym 2,3-diaminotoluene, toluene-2,3-diamine, 3-methyl-o-phenylenediamine, 1,2-benzenediamine, 3-methyl, 3-methyl-1,2-phenylenediamine, 2,3-tolylenediamine, 2,3-toluylenediamine, 1,2-diamino-3-methylbenzene, 3-methyl-1,2-benzenediamine, 1-methyl-2,3-phenylenediamine
PubChem CID 17593
IUPAC Name 3-methylbenzene-1,2-diamine
SMILES CC1=CC=CC(N)=C1N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
AC305670010
View Documents Promotion Details
N/A 1g Glass bottle
Each for $62.32
Only null left
 
AC305670050
View Documents Promotion Details
N/A 5g Glass bottle
Each for $198.52
Only null left
 
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

2687-25-4
122.17
AXNUJYHFQHQZBE-UHFFFAOYSA-N
17593
CC1=CC=CC(N)=C1N
C7H10N2
MFCD00011589
2,3-diaminotoluene, toluene-2,3-diamine, 3-methyl-o-phenylenediamine, 1,2-benzenediamine, 3-methyl, 3-methyl-1,2-phenylenediamine, 2,3-tolylenediamine, 2,3-toluylenediamine, 1,2-diamino-3-methylbenzene, 3-methyl-1,2-benzenediamine, 1-methyl-2,3-phenylenediamine
3-methylbenzene-1,2-diamine

Specifications

2687-25-4
100.0
96% min. (HPLC)
CH3C6H3(NH2)2
2,3-diaminotoluene, toluene-2,3-diamine, 3-methyl-o-phenylenediamine, 1,2-benzenediamine, 3-methyl, 3-methyl-1,2-phenylenediamine, 2,3-tolylenediamine, 2,3-toluylenediamine, 1,2-diamino-3-methylbenzene, 3-methyl-1,2-benzenediamine, 1-methyl-2,3-phenylenediamine
AXNUJYHFQHQZBE-UHFFFAOYSA-N
3-methylbenzene-1,2-diamine
17593
97%
96.0
Authentic
C7H10N2
MFCD00011589
Solubility in water: miscible. Other solubilities: soluble in diethyl ether and ethanol
CC1=CC=CC(N)=C1N
122.17
122.17
2, 3-Diaminotoluene, 97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
May cause an allergic skin reaction.
Causes serious eye irritation.
Suspected of causing genetic defects.
May cause cancer.<b

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fre

GHS Signal Word: Danger

EINECSNumber : 220-248-5

RTECSNumber : XS9550000

TSCA : TSCA

SDS
Documents

Documents

RUO – Research Use Only