missing translation for 'onlineSavingsMsg'
Learn More

2,3-Diamino-6-chloropyridine, 95%

CAS: 40851-95-4 | C5H6ClN3 | 143.57 g/mol

$135.58 - $453.68

Chemical Identifiers

CAS 40851-95-4
Molecular Formula C5H6ClN3
Molecular Weight (g/mol) 143.57
MDL Number MFCD00209966
InChI Key QEIRYIILFUVXAM-UHFFFAOYSA-N
Synonym 2,3-diamino-6-chloropyridine, 6-chloro-2,3-diaminopyridine, 2,3-pyridinediamine,6-chloro, 2,3-pyridinediamine, 6-chloro, 6-chloro-pyridine-2,3-diamine, 6-chloro-2,3-pyridinediamine, pubchem19958, 6-chloro 2,3-pyridindiamine, 2,3-diamino-6-chloro-pyridine
PubChem CID 10630615
IUPAC Name 6-chloropyridine-2,3-diamine
SMILES NC1=C(N)N=C(Cl)C=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6171603
View Documents
Thermo Scientific Chemicals
H6171603
1 g
Each for $135.58
Only null left
 
AAH6171606
View Documents
Thermo Scientific Chemicals
H6171606
5 g
Each for $453.68
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

40851-95-4
143.57
QEIRYIILFUVXAM-UHFFFAOYSA-N
10630615
NC1=C(N)N=C(Cl)C=C1
C5H6ClN3
MFCD00209966
2,3-diamino-6-chloropyridine, 6-chloro-2,3-diaminopyridine, 2,3-pyridinediamine,6-chloro, 2,3-pyridinediamine, 6-chloro, 6-chloro-pyridine-2,3-diamine, 6-chloro-2,3-pyridinediamine, pubchem19958, 6-chloro 2,3-pyridindiamine, 2,3-diamino-6-chloro-pyridine
6-chloropyridine-2,3-diamine

Specifications

40851-95-4
MFCD00209966
UN2811
QEIRYIILFUVXAM-UHFFFAOYSA-N
6-chloropyridine-2,3-diamine
10630615
95%
C5H6ClN3
1 g
2,3-diamino-6-chloropyridine, 6-chloro-2,3-diaminopyridine, 2,3-pyridinediamine,6-chloro, 2,3-pyridinediamine, 6-chloro, 6-chloro-pyridine-2,3-diamine, 6-chloro-2,3-pyridinediamine, pubchem19958, 6-chloro 2,3-pyridindiamine, 2,3-diamino-6-chloro-pyridine
NC1=C(N)N=C(Cl)C=C1
143.57
143.57
2,3-Diamino-6-chloropyridine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Store at -20°C

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only