missing translation for 'onlineSavingsMsg'
Learn More

2,3-Diamino-6-chloropyridine, 95%

Catalog No. AAH6171603
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6171603 1 g
AAH6171606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6171603 Supplier Thermo Scientific Chemicals Supplier No. H6171603
Only null left

CAS: 40851-95-4 | C5H6ClN3 | 143.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 40851-95-4
Molecular Formula C5H6ClN3
Molecular Weight (g/mol) 143.57
MDL Number MFCD00209966
InChI Key QEIRYIILFUVXAM-UHFFFAOYSA-N
Synonym 2,3-diamino-6-chloropyridine, 6-chloro-2,3-diaminopyridine, 2,3-pyridinediamine,6-chloro, 2,3-pyridinediamine, 6-chloro, 6-chloro-pyridine-2,3-diamine, 6-chloro-2,3-pyridinediamine, pubchem19958, 6-chloro 2,3-pyridindiamine, 2,3-diamino-6-chloro-pyridine
PubChem CID 10630615
IUPAC Name 6-chloropyridine-2,3-diamine
SMILES NC1=C(N)N=C(Cl)C=C1

Specifications

Quantity 1 g
UN Number UN2811
Formula Weight 143.57
Percent Purity 95%
Chemical Name or Material 2,3-Diamino-6-chloropyridine
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only