missing translation for 'onlineSavingsMsg'
Learn More

2,3-Diamino-5-chloropyridine, 97%

Catalog No. p-100201
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC350340050 5 g Glass bottle
AC350340250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC350340050 Supplier Thermo Scientific Chemicals Supplier No. 350340050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 25710-20-7 | C5H6ClN3 | 143.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 25710-20-7
Molecular Formula C5H6ClN3
Molecular Weight (g/mol) 143.58
MDL Number MFCD01230986
InChI Key ZRCMCGQDIYNWDX-UHFFFAOYSA-N
Synonym 2,3-diamino-5-chloropyridine, 5-chloro-2,3-diaminopyridine, 5-chloro-2,3-pyridinediamine, 5-chloro-pyridine-2,3-diamine, 2,3-pyridinediamine,5-chloro, pubchem5388, acmc-1cjkt, 5-chloro-2,3diaminopyridine, 5-chloropyridin-2,3-diamine, 2,3-di-amino-5-chloropyridine
PubChem CID 1268254
IUPAC Name 5-chloropyridine-2,3-diamine
SMILES C1=C(C(=NC=C1Cl)N)N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 164°C to 173°C
Color Brown to White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Beilstein 22,II,394
Solubility Information Solubility in water: freely soluble in hot water.,freely soluble in methanol,ethyl acetate,mdc
Formula Weight 143.58
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 2, 3-Diamino-5-chloropyridine, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Harmful in contact with skin.
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
TSCA TSCA

RUO – Research Use Only