missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2',3'-Di-O-acetyl-5'-deoxy-5-fluorocytidine 98.0+%, TCI America™
$106.33 - $848.69
Chemical Identifiers
| CAS | 161599-46-8 |
|---|---|
| Molecular Formula | C13H16FN3O6 |
| Molecular Weight (g/mol) | 329.284 |
| MDL Number | MFCD07369302 |
| InChI Key | NWJBWNIUGNXJGO-RPULLILYSA-N |
| PubChem CID | 11809635 |
| IUPAC Name | [(2R,3R,4R,5R)-4-acetyloxy-5-(4-amino-5-fluoro-2-oxopyrimidin-1-yl)-2-methyloxolan-3-yl] acetate |
| SMILES | CC1C(C(C(O1)N2C=C(C(=NC2=O)N)F)OC(=O)C)OC(=O)C |
Chemical Identifiers
| 161599-46-8 | |
| 329.284 | |
| NWJBWNIUGNXJGO-RPULLILYSA-N | |
| [(2R,3R,4R,5R)-4-acetyloxy-5-(4-amino-5-fluoro-2-oxopyrimidin-1-yl)-2-methyloxolan-3-yl] acetate |
| C13H16FN3O6 | |
| MFCD07369302 | |
| 11809635 | |
| CC1C(C(C(O1)N2C=C(C(=NC2=O)N)F)OC(=O)C)OC(=O)C |
Specifications
| 161599-46-8 | |
| White | |
| MFCD07369302 | |
| NWJBWNIUGNXJGO-RPULLILYSA-N | |
| [(2R,3R,4R,5R)-4-acetyloxy-5-(4-amino-5-fluoro-2-oxopyrimidin-1-yl)-2-methyloxolan-3-yl] acetate | |
| 11809635 | |
| ≥98.0% (HPLC,T) | |
| 2′,3′-Di-O-acetyl-5′-deoxy-5-fluorocytidine |
| 189°C | |
| C13H16FN3O6 | |
| 1 g | |
| CC1C(C(C(O1)N2C=C(C(=NC2=O)N)F)OC(=O)C)OC(=O)C | |
| 329.284 | |
| 329.28 | |
| Crystalline Powder |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator